1-(2-(Trifluoromethyl)pyridin-4-yl)ethanone 97%

Lieferant: Apollo Scientific
Achtung

PC902889-250MG
APOSPC902889-250MGEA 130 EUR
APOSPC902889-250MG
1-(2-(Trifluoromethyl)pyridin-4-yl)ethanone 97%
1-(2-(Trifluoromethyl)pyridin-4-yl)ethanone
Formel: C₈H₆F₃NO
Molecular Weight: 189,135 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11847720
CAS-Nummer: 1060810-86-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR