2-Brom-3,4-difluoranisol 95%

Lieferant: Apollo Scientific

Synonyme: 2-Bromo-3,4-difluoro-1-methoxybenzene

PC902904-25G
APOSPC902904-25GEA 828 EUR
APOSPC902904-25G
2-Brom-3,4-difluoranisol 95%
2-Brom-3,4-difluoranisol
Formel: C₇H₅BrF₂O
Molecular Weight: 223.148 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09261345
CAS-Nummer: 935285-66-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR