3,4-Difluoro-2-methylbenzaldehyde 95%
Lieferant: Apollo Scientific
PC903673-1G
APOSPC903673-1GEA
116
EUR
APOSPC903673-1G
3,4-Difluoro-2-methylbenzaldehyde 95%
3,4-Difluoro-2-methylbenzaldehyde
Formel:
C₈H₆F₂O Molecular Weight: 156,129 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07777151 CAS-Nummer: 847502-84-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...