4-Hydroxy-2-(trifluoromethyl)benzaldehyde

Lieferant: Apollo Scientific
Achtung

PC903683-1G
APOSPC903683-1GEA 26.1 EUR
APOSPC903683-1G
4-Hydroxy-2-(trifluoromethyl)benzaldehyde
4-Hydroxy-2-(trifluoromethyl)benzaldehyde
Formel: C₈H₅F₃O₂
Lagertemperatur: Kühlschrank
MDL: MFCD15528830
CAS-Nummer: 1243395-68-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR