2,3,5,6-Tetrafluoro-4-hydroxybenzaldehyde 95%

Lieferant: Apollo Scientific

PC903690-1G
APOSPC903690-1GEA 342 EUR
APOSPC903690-1G
2,3,5,6-Tetrafluoro-4-hydroxybenzaldehyde 95%
2,3,5,6-Tetrafluoro-4-hydroxybenzaldehyde
Formel: C₇H₂F₄O₂
Molecular Weight: 194,083 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11505943
CAS-Nummer: 24336-73-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR