Potassium 2-methoxyethoxymethyltrifluoroborate 95%

Lieferant: Apollo Scientific
Achtung

PC903705-1G
APOSPC903705-1GEA 229 EUR
APOSPC903705-1G
Potassium 2-methoxyethoxymethyltrifluoroborate 95%
Potassium 2-methoxyethoxymethyltrifluoroborate
Formel: C₄H₉BF₃O₂·K
Lagertemperatur: Gefrierschrank
MDL: MFCD13195184
CAS-Nummer: 910251-13-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR