6-Fluorochromane-2-carboxylic acid 98%

Lieferant: Apollo Scientific

Synonyme: 6-Fluorochroman-2-carboxylic acid, 6-Fluoro-3,4-dihydro-2H-1-benzopyran-2-carboxylic acid

PC903709-25G
APOSPC903709-25GEA 125 EUR
APOSPC903709-25G
6-Fluorochromane-2-carboxylic acid 98%
6-Fluorochromane-2-carboxylic acid
Formel: C₁₀H₉FO₃
Molecular Weight: 196.18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07778357
CAS-Nummer: 99199-60-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR