1-(5-Fluoropyridin-3-yl)ethanone 96%

Lieferant: Apollo Scientific
Achtung

PC903756-100MG PC903756-250MG
APOSPC903756-100MGEA 29 EUR
APOSPC903756-100MG APOSPC903756-250MG
1-(5-Fluoropyridin-3-yl)ethanone 96%
1-(5-Fluoropyridin-3-yl)ethanone
Formel: C₇H₆FNO
Lagertemperatur: Raumtemperatur
MDL: MFCD07375075
CAS-Nummer: 342602-55-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR