4-Chloro-6-fluoro-1H-indole 95+%
Lieferant: Apollo Scientific
|
Achtung
|
PC903779-100MG
PC903779-250MG
APOSPC903779-100MGEA
58
EUR
APOSPC903779-100MG
APOSPC903779-250MG
4-Chloro-6-fluoro-1H-indole 95+%
4-Chloro-6-fluoro-1H-indole
Formel:
C₈H₅ClFN Molecular Weight: 169.583 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD07781557 CAS-Nummer: 885520-79-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...