Methyl 2-cyano-4-fluorobenzoate 97%

Lieferant: Apollo Scientific

PC903820-25G
APOSPC903820-25GEA 134 EUR
APOSPC903820-25G
Methyl 2-cyano-4-fluorobenzoate 97%
Methyl 2-cyano-4-fluorobenzoate
Formel: C₉H₆FNO₂
Molecular Weight: 179,148 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11975805
CAS-Nummer: 127510-96-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR