2-Cyano-4-fluorobenzoic acid 95%

Lieferant: Apollo Scientific

PC903821-1G
APOSPC903821-1GEA 126 EUR
APOSPC903821-1G
2-Cyano-4-fluorobenzoic acid 95%
2-Cyano-4-fluorobenzoic acid
Formel: C₈H₄FNO₂
Molecular Weight: 165,121 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11110402
CAS-Nummer: 1214369-42-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR