2-Fluoro-4-methylpyridine-5-carboxyaldehyde 95%

Lieferant: Apollo Scientific

Synonyme: 6-Fluoro-4-methylnicotinaldehyde

PC903832-1G
APOSPC903832-1GEA 101 EUR
APOSPC903832-1G
2-Fluoro-4-methylpyridine-5-carboxyaldehyde 95%
2-Fluoro-4-methylpyridine-5-carboxyaldehyde
Formel: C₇H₆FNO
Molecular Weight: 139,127 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18447598
CAS-Nummer: 1266253-68-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR