Potassium cyclopentylmethyltrifluoroborate 96%

Lieferant: Apollo Scientific

PC903841-1G PC903841-5G
APOSPC903841-1GEA 0 EUR
APOSPC903841-1G APOSPC903841-5G
Potassium cyclopentylmethyltrifluoroborate 96%
Potassium cyclopentylmethyltrifluoroborate
Formel: C₆H₁₁BF₃·K
Lagertemperatur: Gefrierschrank
MDL: MFCD11052656
CAS-Nummer: 331282-37-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR