Tetraethylammoniumfluorid 98%

Lieferant: Apollo Scientific

PC903855-100G PC903855-25G
APOSPC903855-100GEA 270 EUR
APOSPC903855-100G APOSPC903855-25G
Tetraethylammoniumfluorid 98%
Tetraethylammonium fluoride
Formel: C₈H₂₀FN
Lagertemperatur: Raumtemperatur
MDL: MFCD00011826
CAS-Nummer: 665-46-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR