5-Cyano-2-fluorobenzene-1-sulphonyl chloride 95%

Lieferant: Apollo Scientific
Gefahr

PC903871-1G
APOSPC903871-1GEA 287 EUR
APOSPC903871-1G
5-Cyano-2-fluorobenzene-1-sulphonyl chloride 95%
5-Cyano-2-fluorobenzene-1-sulphonyl chloride
Formel: C₇H₃ClFNO₂S
Molecular Weight: 219,621 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18394026
CAS-Nummer: 1101120-80-2
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR