(1-Methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl)methanol 95%

Lieferant: Apollo Scientific

PC903884-1G
APOSPC903884-1GEA 538 EUR
APOSPC903884-1G
(1-Methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl)methanol 95%
(1-Methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl)methanol
Formel: C₆H₇F₃N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD09040887
CAS-Nummer: 540468-96-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR