2-Bromo-4-fluoro-5-methoxybenzoic acid 95%

Lieferant: Apollo Scientific

PC903899-1G
APOSPC903899-1GEA 1050 EUR
APOSPC903899-1G
2-Bromo-4-fluoro-5-methoxybenzoic acid 95%
2-Bromo-4-fluoro-5-methoxybenzoic acid
Formel: C₈H₆BrFO₃
Molecular Weight: 249,034 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13193242
CAS-Nummer: 1007455-21-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR