(5-Chloro-3-fluoropyridin-2-yl)methanol 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC903907-250MG
APOSPC903907-250MGEA
104
EUR
APOSPC903907-250MG
(5-Chloro-3-fluoropyridin-2-yl)methanol 98%
(5-Chloro-3-fluoropyridin-2-yl)methanol
Formel:
C₆H₅ClFNO Molecular Weight: 161.561 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD16607027 CAS-Nummer: 214055-12-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...