3,5-Difluorphenylisothiocyanat 95%

Lieferant: Apollo Scientific

PC903946-5G
APOSPC903946-5GEA 560 EUR
APOSPC903946-5G
3,5-Difluorphenylisothiocyanat 95%
3,5-Difluorphenylisothiocyanat
Formel: C₇H₃F₂NS
Molecular Weight: 171,17 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 302912-39-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR