2-Chloro-3,4-difluorobenzaldehyde 98%

Lieferant: Apollo Scientific
Achtung

PC903986-250MG
APOSPC903986-250MGEA 59.6 EUR
APOSPC903986-250MG
2-Chloro-3,4-difluorobenzaldehyde 98%
2-Chloro-3,4-difluorobenzaldehyde
Formel: C₇H₃ClF₂O
Molecular Weight: 176,5479 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD18415551
CAS-Nummer: 1261869-07-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR