trans-2-Fluorcyclohexanol 95%

Lieferant: Apollo Scientific

PC903994-1G
APOSPC903994-1GEA 328 EUR
APOSPC903994-1G
trans-2-Fluorcyclohexanol 95%
trans-2-Fluorcyclohexanol
Formel: C₆H₁₁FO
Lagertemperatur: Raumtemperatur
MDL: MFCD08146648
CAS-Nummer: 656-60-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR