5-Fluoro-2-(methylthio)pyrimidine 95%

Lieferant: Apollo Scientific

PC904024-1G
APOSPC904024-1GEA 0 EUR
APOSPC904024-1G
5-Fluoro-2-(methylthio)pyrimidine 95%
5-Fluoro-2-(methylthio)pyrimidine
Formel: C₅H₅FN₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD11858599
CAS-Nummer: 6090-37-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR