3-Chloro-5-(trifluoromethyl)pyridazine

Lieferant: Apollo Scientific

PC904074-1G
APOSPC904074-1GEA 590 EUR
APOSPC904074-1G
3-Chloro-5-(trifluoromethyl)pyridazine
3-Chloro-5-(trifluoromethyl)pyridazine
Formel: C₅H₂ClF₃N₂
Molecular Weight: 182,531 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20275278
CAS-Nummer: 845617-99-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR