N-Isopropyl-3-fluoroaniline 95%

Lieferant: Apollo Scientific

PC904100-1G PC904100-250MG
APOSPC904100-1GEA 0 EUR
APOSPC904100-1G APOSPC904100-250MG
N-Isopropyl-3-fluoroaniline 95%
N-Isopropyl-3-fluoroaniline
Formel: C₉H₁₂FN
Lagertemperatur: Raumtemperatur
MDL: MFCD04972886
CAS-Nummer: 121431-27-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR