3-(Trifluoromethyl)pyridine-4-carboxamide 95%
Lieferant: Apollo Scientific
Synonyme:
3-(Trifluoromethyl)isonicotinamide
PC904151-1G
APOSPC904151-1GEA
53.8
EUR
APOSPC904151-1G
3-(Trifluoromethyl)pyridine-4-carboxamide 95%
3-(Trifluoromethyl)pyridine-4-carboxamide
Formel:
C₇H₅F₃N₂O Molecular Weight: 190,123 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22684694 CAS-Nummer: 1416713-20-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...