Potassium (benzyloxycarbonylamino)methyltrifluoroborate

Lieferant: Apollo Scientific
Achtung

PC904164-1G PC904164-5G
APOSPC904164-1GEA 314 EUR
APOSPC904164-1G APOSPC904164-5G
Potassium (benzyloxycarbonylamino)methyltrifluoroborate
Potassium (benzyloxycarbonylamino)methyltrifluoroborate
Formel: C₉H₁₀BF₃NO₂·K
Molecular Weight: 271.086 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19982304
CAS-Nummer: 1695529-70-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR