2-[(Ethylamino)methyl]-4-fluorophenol 95%

Lieferant: Apollo Scientific

PC904176-1G PC904176-250MG
APOSPC904176-1GEA 435 EUR
APOSPC904176-1G APOSPC904176-250MG
2-[(Ethylamino)methyl]-4-fluorophenol 95%
2-[(Ethylamino)methyl]-4-fluorophenol
Formel: C₉H₁₂FNO
Molecular Weight: 169.196 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21641326
CAS-Nummer: 1363166-27-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR