3,4-Difluorbenzoylacetonitril 95+%

Lieferant: Apollo Scientific

Synonyme: 3-(3,4-Difluorophenyl)-3-oxopropanenitrile

PC9042-1G
APOSPC9042-1GEA 78.3 EUR
APOSPC9042-1G
3,4-Difluorbenzoylacetonitril 95+%
3,4-Difluorbenzoylacetonitril
Formel: C₉H₅F₂NO
Molecular Weight: 181.14
Lagertemperatur: Raumtemperatur
CAS-Nummer: 71682-97-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR