Potassium 2-(tetrahydro-2h-pyran-2-yloxy)ethyltrifluoroborate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Potassium trifluoro-[2-(oxan-2-yloxy)ethyl]boranuide, Potassium trifluoro(2-((tetrahydro-2H-pyran-2-yl)oxy)ethyl)borate
PC904235-1G
PC904235-250MG
PC904235-5G
APOSPC904235-1GEA
89.9
EUR
APOSPC904235-1G
APOSPC904235-250MG
APOSPC904235-5G
Potassium 2-(tetrahydro-2h-pyran-2-yloxy)ethyltrifluoroborate
Potassium 2-(tetrahydro-2h-pyran-2-yloxy)ethyltrifluoroborate
Formel:
C₇H₁₃BF₃O₂·K Molecular Weight: 236.081 g/mol Schmelzpunkt: 182…185 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD22684702 CAS-Nummer: 1408168-76-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...