Potassium (2-benzyloxyethyl)trifluoroborate 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Potassium (2-(benzyloxy)ethyl)trifluoroborate
PC904250-1G
APOSPC904250-1GEA
161
EUR
APOSPC904250-1G
Potassium (2-benzyloxyethyl)trifluoroborate 95%
Potassium (2-benzyloxyethyl)trifluoroborate
Formel:
C₉H₁₁BF₃O·K Molecular Weight: 242,088 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD22684708 CAS-Nummer: 1408168-73-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...