Methyl 2-fluoro-5-hydroxy-4-methylbenzoate 97%

Lieferant: Apollo Scientific

PC904299-5G
APOSPC904299-5GEA 1850 EUR
APOSPC904299-5G
Methyl 2-fluoro-5-hydroxy-4-methylbenzoate 97%
Methyl 2-fluoro-5-hydroxy-4-methylbenzoate
Formel: C₉H₉FO₃
Molecular Weight: 184,164 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19440272
CAS-Nummer: 1378655-77-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR