2-Fluoro-5-hydroxymethyl-4-methylpyridine 95%

Lieferant: Apollo Scientific

PC904307-1G
APOSPC904307-1GEA 538 EUR
APOSPC904307-1G
2-Fluoro-5-hydroxymethyl-4-methylpyridine 95%
2-Fluoro-5-hydroxymethyl-4-methylpyridine
Formel: C₇H₈FNO
Molecular Weight: 141,143 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22392285
CAS-Nummer: 1394899-05-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR