2-Chloro-1-(5-fluoro-2-pyridyl)ethanone 95%

Lieferant: Apollo Scientific

PC904365-1G
APOSPC904365-1GEA 632 EUR
APOSPC904365-1G
2-Chloro-1-(5-fluoro-2-pyridyl)ethanone 95%
2-Chloro-1-(5-fluoro-2-pyridyl)ethanone
Formel: C₇H₅ClFNO
Lagertemperatur: Raumtemperatur
MDL: MFCD17169899
CAS-Nummer: 1104606-44-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR