5-Fluorobenzoxazole-2-thiol 98%

Lieferant: Apollo Scientific

Synonyme: 5-Fluorobenzo[d]oxazole-2-thiol

PC904486-5G
APOSPC904486-5GEA 60.9 EUR
APOSPC904486-5G
5-Fluorobenzoxazole-2-thiol 98%
5-Fluorobenzoxazole-2-thiol
Formel: C₇H₄FNOS
Molecular Weight: 169.18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08447021
CAS-Nummer: 13451-78-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR