6-(Trifluoromethyl)pyridazin-3(2H)-one 95%

Lieferant: Apollo Scientific

PC904495-1G
APOSPC904495-1GEA 106 EUR
APOSPC904495-1G
6-(Trifluoromethyl)pyridazin-3(2H)-one 95%
6-(Trifluoromethyl)pyridazin-3(2H)-one
Formel: C5H3F3N2O
Molecular Weight: 164.0853 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09754152
CAS-Nummer: 174607-36-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR