2,4-Bis(trifluormethyl)phenylacetonitril 98%

Lieferant: Apollo Scientific
Achtung

PC9045-1G PC9045-5G
APOSPC9045-1GEA 39.3 EUR
APOSPC9045-1G APOSPC9045-5G
2,4-Bis(trifluormethyl)phenylacetonitril 98%
2,4-Bis(trifluormethyl)phenylacetonitril
Formel: C₁₀H₅F₆N
Lagertemperatur: Raumtemperatur
CAS-Nummer: 201789-28-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR