2-(5-Fluoropyridin-3-yl)acetic acid 98%

Lieferant: Apollo Scientific

PC904503-1G PC904503-250MG
APOSPC904503-1GEA 369 EUR
APOSPC904503-1G APOSPC904503-250MG
2-(5-Fluoropyridin-3-yl)acetic acid 98%
2-(5-Fluoropyridin-3-yl)acetic acid
Formel: C₇H₆FNO₂
Molecular Weight: 155.13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09924977
CAS-Nummer: 38129-24-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR