5-(4-Fluorophenyl)isoxazole-3-carboxaldehyde 95%
Lieferant: Apollo Scientific
PC904563-1G
APOSPC904563-1GEA
241
EUR
APOSPC904563-1G
5-(4-Fluorophenyl)isoxazole-3-carboxaldehyde 95%
5-(4-Fluorophenyl)isoxazole-3-carboxaldehyde
Formel:
C₁₀H₆FNO₂ Molecular Weight: 191,159 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06798099 CAS-Nummer: 640292-06-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...