3,6-Dichloro-4-fluoropyridazine

Lieferant: Apollo Scientific
Achtung

PC904584-1G
APOSPC904584-1GEA 184 EUR
APOSPC904584-1G
3,6-Dichloro-4-fluoropyridazine
3,6-Dichloro-4-fluoropyridazine
Formel: C₄HCl₂FN₂
Molecular Weight: 166,969 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21648467
CAS-Nummer: 92920-33-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR