Tetrahydro-1-(phenylmethyl)thiophenium hexafluorophosphate 95%

Lieferant: Apollo Scientific
Achtung

PC904661-5G
APOSPC904661-5GEA 197 EUR
APOSPC904661-5G
Tetrahydro-1-(phenylmethyl)thiophenium hexafluorophosphate 95%
Tetrahydro-1-(phenylmethyl)thiophenium hexafluorophosphate
Formel: C₁₁H₁₅F₆PS
Molecular Weight: 324,27 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD22421760
CAS-Nummer: 87301-53-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR