2,5-Difluoro-4-methylaniline 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC904700-1G
APOSPC904700-1GEA
29
EUR
APOSPC904700-1G
2,5-Difluoro-4-methylaniline 97%
2,5-Difluoro-4-methylaniline
Formel:
C₇H₇F₂N Molecular Weight: 143,134 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11519027 CAS-Nummer: 878285-14-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...