6-Fluoro-1-methyl-1H-indole-3-carbaldehyde 95%

Lieferant: Apollo Scientific

PC904777-1G
APOSPC904777-1GEA 112 EUR
APOSPC904777-1G
6-Fluoro-1-methyl-1H-indole-3-carbaldehyde 95%
6-Fluoro-1-methyl-1H-indole-3-carbaldehyde
Formel: C₁₀H₈FNO
Molecular Weight: 177.18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07790764
CAS-Nummer: 441715-93-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR