3-Chloro-6-fluorobenzo[d]isoxazole 97%

Lieferant: Apollo Scientific
Achtung

PC904835-250MG
APOSPC904835-250MGEA 112 EUR
APOSPC904835-250MG
3-Chloro-6-fluorobenzo[d]isoxazole 97%
3-Chloro-6-fluorobenzo[d]isoxazole
Formel: C₇H₃ClFNO
Molecular Weight: 171,556 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD16996105
CAS-Nummer: 374554-89-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR