4-Fluor-7-methylisatin 95%

Lieferant: Apollo Scientific

PC904862-1G
APOSPC904862-1GEA 357 EUR
APOSPC904862-1G
4-Fluor-7-methylisatin 95%
4-Fluor-7-methylisatin
Formel: C₉H₆FNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD06409071
CAS-Nummer: 668-24-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR