2-(Trifluoroacetyl)cyclohexanone

Lieferant: Apollo Scientific
Achtung

PC904873-1G PC904873-5G
APOSPC904873-1GEA 34.8 EUR
APOSPC904873-1G APOSPC904873-5G
2-(Trifluoroacetyl)cyclohexanone
2-(Trifluoroacetyl)cyclohexanone
Formel: C₈H₉F₃O₂
Molecular Weight: 194.151 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07779548
CAS-Nummer: 387-89-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR