1-(5-Fluoropyridin-2-yl)ethanamine 95%

Lieferant: Apollo Scientific

PC904892-1G PC904892-250MG
APOSPC904892-1GEA 0 EUR
APOSPC904892-1G APOSPC904892-250MG
1-(5-Fluoropyridin-2-yl)ethanamine 95%
1-(5-Fluoropyridin-2-yl)ethanamine
Formel: C₇H₉FN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD18260255
CAS-Nummer: 915720-57-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR