1-(2,2,2-Trifluoroacetyl)-3-azetidinecarboxylic acid 95%

Lieferant: Apollo Scientific

PC904931-1G PC904931-5G
APOSPC904931-1GEA 132 EUR
APOSPC904931-1G APOSPC904931-5G
1-(2,2,2-Trifluoroacetyl)-3-azetidinecarboxylic acid 95%
1-(2,2,2-Trifluoroacetyl)-3-azetidinecarboxylic acid
Formel: C₆H₆F₃NO₃
Molecular Weight: 197.11 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22421758
CAS-Nummer: 1001026-41-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR