(2-Bromethyl)(diphenyl)sulfoniumtrifluormethansulfonat 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: (2-Bromoethyl)diphenylsulphonium Triflate

PC904948-5G
APOSPC904948-5GEA 126 EUR
APOSPC904948-5G
(2-Bromethyl)(diphenyl)sulfoniumtrifluormethansulfonat 98%
(2-Bromoethyl)diphenylsulphonium trifluoromethanesulphonate
Formel: C₁₅H₁₄BrF₃O₃S₂
Molecular Weight: 443.29 g/mol
Schmelzpunkt: 85…90 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD12546049
CAS-Nummer: 247129-85-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR