2-(4-Fluorophenyl)azepane 95%
Lieferant: Apollo Scientific
PC904975-1G
APOSPC904975-1GEA
0
EUR
APOSPC904975-1G
2-(4-Fluorophenyl)azepane 95%
2-(4-Fluorophenyl)azepane
Formel:
C₁₂H₁₆FN Lagertemperatur: Raumtemperatur |
MDL:
MFCD02663706 CAS-Nummer: 168890-44-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...