2-Ethoxy-5-fluoropyrimidin-4-amine 95%

Lieferant: Apollo Scientific

PC905063-1G
APOSPC905063-1GEA 74.1 EUR
APOSPC905063-1G
2-Ethoxy-5-fluoropyrimidin-4-amine 95%
2-Ethoxy-5-fluoropyrimidin-4-amine
Formel: C₆H₈FN₃O
Molecular Weight: 157,146 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18973123
CAS-Nummer: 56076-21-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR